Skip to Content
Merck
All Photos(1)

Key Documents

24-2109

Sigma-Aldrich

3-Methyl-1-phenyl-2-pyrazoline-5-one

Synonym(s):

5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
Beilstein:
609575
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
application(s):
peptide synthesis
bp:
287 °C/265 mmHg (lit.)
Pricing and availability is not currently available.

form

solid

availability

available only in Japan

dilution

(for analytical testing)

bp

287 °C/265 mmHg (lit.)

mp

126-128 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC1=NN(C(=O)C1)c2ccccc2

InChI

1S/C10H10N2O/c1-8-7-10(13)12(11-8)9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QELUYTUMUWHWMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

environmental analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Monturiol-Gross et al.
The Journal of infectious diseases, 206(8), 1218-1226 (2012-08-21)
Clostridium perfringens, the most broadly distributed pathogen in nature, produces a prototype phospholipase C, also called α-toxin, which plays a key role in the pathogenesis of gas gangrene. α-Toxin causes plasma membrane disruption at high concentrations, but the role of
Manabu Ohta et al.
Brain research, 1490, 184-192 (2012-09-18)
Traumatic axonal injury (TAI), a feature of traumatic brain injury (TBI), progressively evolves over hours through impaired axonal transport and is thought to be a major contributor to cognitive dysfunction. In spite of various studies suggesting that pharmacologic or physiologic
Ajmal Ahmad et al.
Molecular and cellular biochemistry, 367(1-2), 215-225 (2012-06-01)
Stroke is a life-threatening disease with major cause of mortality and morbidity worldwide. The neuronal damage following cerebral ischemia is a serious risk to stroke patients. Oxidative stress and apoptotic damage play an important role in cerebral ischemic pathogenesis and
Mitsugi Shimoda et al.
World journal of gastroenterology, 18(27), 3520-3526 (2012-07-25)
To investigate the effect of E3-methyl-1-phenyl-2-pyrazolin-5-one (Edr) on hepatic ischemia-reperfusion (I/R) injury and liver regeneration in a porcine hepatectomy model. One hour ischemia was induced by occluding the vessels and the bile duct of the right and median lobes. A
Guibin Wang et al.
Neuroscience letters, 543, 72-77 (2013-04-09)
Edaravone, a free radical scavenger, is the first clinical drug of neuroprotection for ischemic stroke patients in the world, and has been shown to be an effective agent to alleviate cerebral ischemic injury. It has been established that acidosis is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service