Skip to Content
Merck
All Photos(3)

Key Documents

M70800

Sigma-Aldrich

3-Methyl-1-phenyl-2-pyrazoline-5-one

99%, for peptide synthesis

Synonym(s):

Edaravone, 5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$80.80
100 G
$82.70
500 G
$176.00

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$80.80
100 G
$82.70
500 G
$176.00

About This Item

Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
Beilstein:
609575
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%

Assay

99%

form

powder, crystals or chunks

bp

287 °C/265 mmHg (lit.)

mp

126-128 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC1=NN(C(=O)C1)c2ccccc2

InChI

1S/C10H10N2O/c1-8-7-10(13)12(11-8)9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QELUYTUMUWHWMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-1-phenyl-2-pyrazoline-5-one may be used:
  • As a reagent for the detection of reducing carbohydrates by ESI/MALDI-MS.[1]
  • To improve the sensitivity of reducing mono- and oligo-saccharides for their subsequent determination using capillary zone electrophoresis.[2]
  • For the precolumn derivatization of the monosaccharide components of Salvia miltiorrhiza, liguspyragine hydrochloride, and glucose injection prior to their analysis by high performance liquid chromatography (HPLC).[3]

Reagent for detection of reducing carbohydrates by ESI/MALDI -MS.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capillary zone electrophoresis of reducing mono-and oligo-saccharides as the borate complexes of their 3-methyl-1-phenyl-2-pyrazolin-5-one derivatives
Honda S, et al.
Carbohydrate Research, 215(1), 193-198 (1991)
Fa-Huan Ge et al.
Journal of analytical methods in chemistry, 2017, 9245620-9245620 (2017-05-11)
Salvia miltiorrhiza, liguspyragine hydrochloride, and glucose injection (SLGI) was made of Salvia miltiorrhiza Bge., liguspyragine hydrochloride, glucose, and glycerin. There were many kinds of monosaccharide components in SLGI, which might be from the raw material and Salvia miltiorrhiza Bge. Separation
Influence of the labeling group on ionization and fragmentation of carbohydrates in mass spectrometry
Lattova E, et al.
Journal of the American Society For Mass Spectrometry, 16(5), 683-696 (2005)
Xian-Wen Yang et al.
Journal of natural products, 70(4), 521-525 (2007-03-09)
Six new mixed lignan-neolignans and 20 known compounds were isolated from the whole plant of Tarenna attenuata. By analysis of physical and spectroscopic data, the structures of the new compounds were elucidated as (1R,5R,6R)-6-{4-O-[2-(1-(4-hydroxy-3-methoxyphenyl))glycerol]-3,5-dimethoxyphenyl}-3,7-dioxabicyclo[3.3.0]octan-2-one (1), 5' '-methoxyhedyotisol A (2), 4'
Hidehiko Nakagawa et al.
Bioorganic & medicinal chemistry letters, 16(23), 5939-5942 (2006-09-26)
We synthesized various 3-methyl-1-phenyl-5-pyrazolone (edaravone) derivatives and evaluated their oxidation potential and hydroxyl radical scavenging activity. It was found 3-methyl-1-(pyridin-2-yl)-5-pyrazolone had a much higher ability to scavenge the radical than did edaravone itself. Its efficient radical scavenging activity was assumed

Articles

Glycans play a key role in protein structure and disease; representation on cell surfaces is the glycome.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service