Skip to Content
Merck
All Photos(1)

Documents

24-0970

Sigma-Aldrich

Peptone from animal tissue

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41100000

biological source

animal

grade

SAJ first grade

form

solid

availability

available only in Japan

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saoussen Turki et al.
Bioresource technology, 100(10), 2724-2731 (2009-02-04)
In this work the effect of several organic nitrogen sources on lipase production in Yarrowia lipolytica LgX64.81 overproducing mutant was studied. Among them, tryptone and peptone showed the most prominent stimulatory effect. Interestingly, only tryptic and peptic casein digest were
M Taskin et al.
Journal of applied microbiology, 111(4), 826-834 (2011-07-19)
Peptones are one of the most expensive constituents of microbial media. This study was undertaken to prepare the peptone from waste chicken feathers through a new process. The chemical analysis of chicken feather peptone (CFP) was performed. The ability of
Aiqi Fang et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(32), 13225-13229 (2009-08-05)
In the hope of developing a vaccine against Clostridium difficile based on its toxin(s), we have developed a fermentation medium for the bacterium that results in the formation of Toxin A and contains no meat or dairy products, thus obviating
Mesut Taskin et al.
International journal of food sciences and nutrition, 63(5), 597-602 (2011-12-06)
This study was performed to investigate the usability of chicken feather hydrolysate (Chicken feather peptone (CFP)) as substrate for mycelial biomass and extracellular polysaccharides (EPS) production from edible mushroom Morchella esculenta. The ability of CFP to support biomass and EPS
Paulraj Kanmani et al.
Preparative biochemistry & biotechnology, 41(1), 40-52 (2011-01-14)
Statistics-based experimental designs were used to develop a cost-effective medium for enhanced production of viable cells and bacteriocin by probiotic Enterococcus faecium MC13. Carbon, nitrogen, and mineral sources were first screened by one-variable-at-a-time (OVAT) methods. In order to increase yield

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service