Skip to Content
Merck
All Photos(1)

Key Documents

23-0510

Sigma-Aldrich

Soybean oil

CP

Synonym(s):

Soya oil from Glycine max

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

grade

CP

form

oil

availability

available only in Japan

refractive index

n20/D 1.4743 (lit.)

density

0.917 g/mL at 25 °C (lit.)

functional group

carboxylic acid
oleic acid

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela M Nivala et al.
Metabolism: clinical and experimental, 62(5), 753-760 (2013-01-15)
In cell systems, saturated fatty acids, compared to unsaturated fatty acids, induce a greater degree of ER stress and inflammatory signaling in a number of cell types, including hepatocytes and adipocytes. The aim of the present study was to determine
Li Liu et al.
Biochimica et biophysica acta, 1830(6), 3447-3453 (2013-02-12)
Superparamagnetic iron-oxide nanoparticles are useful as contrast agents for anatomical, functional and cellular MRI, drug delivery agents, and diagnostic biosensors. Nanoparticles are generally cleared by the reticuloendothelial system (RES), in particular taken up by Kupffer cells in the liver, limiting
Susana P Alves et al.
PloS one, 8(3), e58386-e58386 (2013-03-14)
Lipid metabolism in the rumen is responsible for the complex fatty acid profile of rumen outflow compared with the dietary fatty acid composition, contributing to the lipid profile of ruminant products. A method for the detailed dimethylacetal and fatty acid
Kara S Hughan et al.
The Journal of clinical endocrinology and metabolism, 98(5), 2062-2069 (2013-03-26)
Overweight/obese (OW/OB) African American (AA) adolescents have a more diabetogenic insulin secretion/sensitivity pattern compared with their American white (AW) peers. The present study investigated β-cell lipotoxicity to test whether increased free fatty acid (FFA) levels result in greater β-cell dysfunction
Management of near fatal mixed tricyclic antidepressant and selective serotonin reuptake inhibitor overdose with Intralipid®20% emulsion.
A Nair et al.
Anaesthesia and intensive care, 41(2), 264-265 (2013-03-28)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service