Skip to Content
Merck
All Photos(1)

Key Documents

19-2835

Sigma-Aldrich

tert-Butyl methyl ether

SAJ special grade, ≥99.0%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
grade:
SAJ special grade
Assay:
≥99.0%
bp:
55-56 °C (lit.)
vapor pressure:
4.05 psi
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

3.1 (vs air)

vapor pressure

4.05 psi

Assay

≥99.0%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

availability

available only in Japan

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

mp

-110 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva M Seeger et al.
Water research, 47(2), 769-780 (2012-12-04)
For several pilot-scale constructed wetlands (CWs: a planted and unplanted gravel filter) and a hydroponic plant root mat (operating at two water levels), used for treating groundwater contaminated with BTEX, the fuel additive MTBE and ammonium, the hydrodynamic behavior was
Torsten C Schmidt et al.
Journal of contaminant hydrology, 70(3-4), 173-203 (2004-05-12)
The fate of fuel oxygenates such as methyl tert-butyl ether (MTBE) in the subsurface is governed by their degradability under various redox conditions. The key intermediate in degradation of MTBE and ethyl tert-butyl ether (ETBE) is tert-butyl alcohol (TBA) which
Sarah K Abbott et al.
Lipids, 48(3), 307-318 (2013-01-29)
We have developed a protocol suitable for high-throughput lipidomic analysis of human brain samples. The traditional Folch extraction (using chloroform and glass-glass homogenization) was compared to a high-throughput method combining methyl-tert-butyl ether (MTBE) extraction with mechanical homogenization utilizing ceramic beads.
Manuela Nagel et al.
Plant, cell & environment, 38(6), 1011-1022 (2014-10-21)
Globally, over 7.4 million accessions of crop seeds are stored in gene banks, and conservation of genotypic variation is pivotal for breeding. We combined genetic and biochemical approaches to obtain a broad overview of factors that influence seed storability and
Ceinwen A Schreiner et al.
Regulatory toxicology and pharmacology : RTP, 70(2 Suppl), S29-S34 (2014-05-24)
Micronucleus and sister chromatid exchange (SCE) tests were performed for vapor condensate of baseline gasoline (BGVC), or gasoline with oxygenates, methyl tert-butyl ether (G/MTBE), ethyl tert butyl ether (G/ETBE), t-amyl methyl ether (G/TAME), diisopropyl ether (G/DIPE), t-butyl alcohol (TBA), or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service