tert-Butyl methyl ether
Synonym(s):
MTBE, Methyl tert-butyl ether, tert-Butyl methyl ether, tert-Butyl methyl ether
Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing