Skip to Content
Merck
All Photos(1)

Key Documents

19-1870

Sigma-Aldrich

Mercury standard solution

suitable for atomic absorption spectrometry, 1 mg/mL Hg, 1000 ppm Hg

Synonym(s):

Mercury(II) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

form

liquid

availability

available only in Japan

concentration

1 mg/mL Hg
1000 ppm Hg

suitability

suitable for atomic absorption spectrometry

SMILES string

[Hg]

InChI

1S/Hg

InChI key

QSHDDOUJBYECFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neelam Yadav et al.
The Indian journal of medical research, 136(2), 211-220 (2012-09-11)
Acute tubular necrosis (ATN) caused by renal ischaemia, renal hypo-perfusion, or nephrotoxic substances is the most common form of acute kidney injury (AKI). There are a few treatment options for this life-threatening disease and the mortality rate exceeds 50 per
Andrés Cruz Hernández et al.
Metallomics : integrated biometal science, 5(4), 398-403 (2013-03-27)
RNA processing is an essential pathway in the regulation of genetic expression in the cell. In this work, Bacillus subtilis was used to understand the effects of mercury on the mechanism of tRNA metabolism. The CVAAS (cold vapor atomic absorption
Sai Jin Xiao et al.
The journal of physical chemistry. B, 116(32), 9565-9569 (2012-07-25)
Though rapid tests were developed for mass screening of prion diseases in the last century, bovine spongiform encephalopathy (BSE) was still epidemic in some European countries. The main reason is that the sensitivity of such tests is insufficient for detecting
Zuzana Knazicka et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(3), 348-353 (2012-12-19)
In this study the NCI-H295R human adrenocortical carcinoma cell line was used as an in vitro biological model to study the effect of mercury (HgCl(2)) on the steroidogenesis. The cells were cultured for 48 h with addition of 1.0; 5.0; 25;
Alexey Vorobev et al.
Applied and environmental microbiology, 79(19), 5918-5926 (2013-07-23)
Many methanotrophs have been shown to synthesize methanobactin, a novel biogenic copper-chelating agent or chalkophore. Methanobactin binds copper via two heterocyclic rings with associated enethiol groups. The structure of methanobactin suggests that it can bind other metals, including mercury. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service