Skip to Content
Merck
All Photos(1)

Key Documents

89459

Supelco

Mercury Standard for ICP

TraceCERT®, 10 mg/L Hg in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$275.00

$275.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$275.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$275.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

concentration

10 mg/L Hg in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Mercury Standard for ICP may be used as a calibration standard for the determination of total mercury by inductively coupled plasma mass spectrometry method (ICP-MS).[1]

Analysis Note

10 mg/L Hg in 12% nitric acid, prepared with high-purity mercury metal, HNO3, and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective organomercury determination by ICP-MS made easy
Spanu D, et al.
Analytica Chimica Acta, 1206, 339553-339553 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service