13-1160
L-Histidine
SAJ special grade, ≥98.5%
Synonym(s):
(S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
84088
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Recommended Products
grade
SAJ special grade
Assay
≥98.5%
form
solid
availability
available only in Japan
mp
282 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
N[C@@H](Cc1c[nH]cn1)C(O)=O
InChI
1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1
InChI key
HNDVDQJCIGZPNO-YFKPBYRVSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Precursor of histamine by action of histidine decarboxylase.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pawel Swietach et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), E2064-E2073 (2013-05-17)
Ca(2+) signaling regulates cell function. This is subject to modulation by H(+) ions that are universal end-products of metabolism. Due to slow diffusion and common buffers, changes in cytoplasmic [Ca(2+)] ([Ca(2+)]i) or [H(+)] ([H(+)]i) can become compartmentalized, leading potentially to
Shweta Singh et al.
Microbial cell factories, 7, 28-28 (2008-10-14)
The large-scale production of G-protein coupled receptors (GPCRs) for functional and structural studies remains a challenge. Recent successes have been made in the expression of a range of GPCRs using Pichia pastoris as an expression host. P. pastoris has a
H M Golder et al.
Journal of dairy science, 97(8), 5131-5150 (2014-06-03)
Ruminal bacterial community composition (BCC) and its associations with ruminal fermentation measures were studied in dairy heifers challenged with combinations of grain, fructose, and histidine in a partial factorial study. Holstein-Friesian heifers (n=30) were randomly allocated to 5 triticale grain-based
Urszula Kosinska Eriksson et al.
Science (New York, N.Y.), 340(6138), 1346-1349 (2013-06-15)
Aquaporins are membrane channels that facilitate the flow of water across biological membranes. Two conserved regions are central for selective function: the dual asparagine-proline-alanine (NPA) aquaporin signature motif and the aromatic and arginine selectivity filter (SF). Here, we present the
Giorgio Zoppellaro et al.
Biopolymers, 91(12), 1064-1082 (2009-06-19)
Six-coordinated heme groups are involved in a large variety of electron transfer reactions because of their ability to exist in both the ferrous (Fe(2+)) and ferric (Fe(3+)) state without any large differences in structure. Our studies on hemes coordinated by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service