Skip to Content
Merck
All Photos(1)

Key Documents

176680

Sigma-Aldrich

Hypophosphorous acid-d3 solution

50 wt. % in D2O, 98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$531.00

$531.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 G
$531.00

About This Item

Linear Formula:
D3PO2
CAS Number:
Molecular Weight:
69.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$531.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

concentration

50 wt. % in D2O

technique(s)

NMR: suitable

density

1.350 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]OP([2H])O[2H]

InChI

1S/H3O2P/c1-3-2/h1-3H/i1D,2D,3D

InChI key

XRBCRPZXSCBRTK-CBYSEHNBSA-N

General description

The product is 50wt.% solution of hypophosphorous acid-d3 in D2O, having an isotopic purity of 98atom%. It can be obtained by proton-deuterium-exchange reaction in D2O. Deuterated hypophosphorous acid can be employed in neutron diffraction studies.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reinvestigation of Crystal Structure and Non-Stoichiometry in Copper Hydride, CuH1-x(0= x= 0.26).
Auer H and Kohlmann H.
Zeitschrift fur Anorganische und Allgemeine Chemie, 640(15), 3159-3165 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service