11-0475
Fluoride standard solution
1 mg/mL F-
Synonym(s):
Sodium fluoride solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
availability
available only in Japan
concentration
1 mg/mL F-
density
1.005 g/mL at 25 °C (lit.)
SMILES string
[F-].[Na+]
InChI
1S/FH.Na/h1H;/q;+1/p-1
InChI key
PUZPDOWCWNUUKD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of endocrinology, 218(1), 99-103 (2013-05-11)
Daily intake of water with fluoride concentrations >1.5 mg/l produces insulin resistance (IR). On the other hand, physical activity increases insulin sensitivity in the muscle. Therefore, the aim of this study was to evaluate the effect of physical activity on IR
Toxicology, 308, 138-145 (2013-04-13)
Sodium fluoride (NaF) is widely used for the treatment of dental caries and dentin hypersensitivity. However, its pro-apoptotic effect on odontoblasts may lead to harmful side-effects. The purpose of this study was to evaluate the pro-apoptotic effects of NaF in
Anesthesiology, 118(5), 1076-1085 (2013-01-29)
Tyrosine (Tyr) kinases and mitogen-activated protein kinases have been thought to participate in the contractile response in various smooth muscles. The aim of the current study was to investigate the involvement of the Tyr kinase pathway in the contraction of
The Journal of clinical dentistry, 24(1), 20-24 (2013-04-04)
The purpose of this study was to determine the viability of dental plaque at 12 hours following brushing with Colgate Great Regular Flavor (0.83% MFP), Crest Pro-Health (0.454% SnF), and Colgate Total Clean Mint Toothpaste (0.3% triclosan/ copolymer/0.22% NaF). Thirty-two
Noise & health, 15(63), 143-147 (2013-04-11)
Bone remodeling disorders have been identified as potential candidates for the etiology underlying some forms of otologic disease. Here, four clinical cases are presented to demonstrate a novel approach to tinnitus management that focuses on two fundamental etiologic bases. First
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service