Merck
  • Sodium fluoride induces apoptosis in odontoblasts via a JNK-dependent mechanism.
Sodium fluoride induces apoptosis in odontoblasts via a JNK-dependent mechanism.
Toxicology (2013-04-13)
Peng Li, Yunpeng Xue, Wenbin Zhang, Fei Teng, Yong Sun, Tiejun Qu, Xingxing Chen, Xiaogang Cheng, Bing Song, Wenjing Luo, Qing Yu
ABSTRACT
Sodium fluoride (NaF) is widely used for the treatment of dental caries and dentin hypersensitivity. However, its pro-apoptotic effect on odontoblasts may lead to harmful side-effects. The purpose of this study was to evaluate the pro-apoptotic effects of NaF in odontoblasts and elucidate the possible underlying molecular mechanisms. NaF generated cytotoxic effects in odontoblast-lineage cell (OLC) in a dose- and time-dependent manner. Exposure of cells to 4mM NaF for 24h induced caspase-3 activation, ultrastructural alterations, and resulted in the translocation of Bax to the mitochondria and the release of cytochrome c from the mitochondrial inter-membrane space into the cytosol, indicating that fluoride-mediated apoptosis is mitochondria-dependent. Fluoride treatment also increased phosphorylation of JNK and ERK, but not p38, and apoptosis induced by fluoride was notably or partly suppressed by treatment with JNK or ERK inhibitors, respectively. Taken together, these findings suggest that NaF induces apoptosis in OLC odontoblasts through a JNK-dependent mitochondrial pathway.
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%