Skip to Content
Merck
All Photos(1)

Key Documents

05-6220

Sigma-Aldrich

Copper(II) sulfate pentahydrate

SAJ first grade, ≥99.0%

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

7.3 mmHg ( 25 °C)

Assay

≥99.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

pH

3.7-4.5 (25 °C, 50 g/L)

mp

110 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela M B Haverroth et al.
Ecotoxicology and environmental safety, 122, 440-447 (2015-09-20)
Copper is a heavy metal found at relatively high concentrations in surface waters around the world. Copper is a micronutrient at low concentrations and is essential to several organisms. At higher concentrations copper can become toxic, which reveal the importance
Bhagwan S Batule et al.
International journal of nanomedicine, 10 Spec Iss, 137-142 (2015-09-09)
We developed a simple but efficient method to synthesize protein-inorganic hybrid nanostructures with a flower-like shape (nanoflowers), which relies on sonication to facilitate the synthesis of the nanoflowers. With this technique, we synthesized nanoflowers containing laccase as a model protein
Fange Liu et al.
The Journal of biological chemistry, 290(25), 15621-15634 (2015-04-29)
The rubredoxin motif is present in over 74,000 protein sequences and 2,000 structures, but few have known functions. A secondary, non-catalytic, rubredoxin-like iron site is conserved in 3-hydroxyanthranilate 3,4-dioxygenase (HAO), from single cellular sources but not multicellular sources. Through the
Florence Haudin et al.
Physical chemistry chemical physics : PCCP, 17(19), 12804-12811 (2015-04-25)
The growth of chemical gardens is studied experimentally in a horizontal confined geometry when a solution of metallic salt is injected into an alkaline solution at a fixed flow rate. Various precipitate patterns are observed-spirals, flowers, worms or filaments-depending on
Anna Golunova et al.
Biomacromolecules, 16(4), 1146-1156 (2015-03-03)
The ability to tailor mechanical properties and architecture is crucial in creating macroporous hydrogel scaffolds for tissue engineering. In the present work, a technique for the modification of the pore size and stiffness of acrylamide-based cryogels is demonstrated via the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service