Skip to Content
Merck
Copper(II) sulfate pentahydrate purum p.a., crystallized, ≥99.0% (RT)

Copper(II) sulfate pentahydrate

Copper(II) sulfate pentahydrate purum p.a., crystallized, ≥99.0% (RT)
Synonym(s):
Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate, Copper(II) sulfate pentahydrate
Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Technique