The dimensions for the 5-liter pressure vessel are 22.9 cm in diameter and 24.1 cm in height (9.0 in. × 9.5 in.), and for the 1-gallon vessel, 22.9 cm in diameter and 21.6 cm in height (9.0 in. × 8.5 in.). User Guides can be provided upon request or downloaded from the website. Drawings are considered confidential and are not shared.
XX6700P01
Dispensing Pressure Vessel, 1 gal
Synonym(s):
Pressure vessel
About This Item
Recommended Products
material
316 stainless steel body
fluoroelastomer O-ring
fluoroelastomer gasket
molded neoprene base
stainless steel fittings
Quality Level
parameter
6.9 bar max. inlet pressure (100 psi)
H
229 mm
~9 in.
I.D.
6.3 mm (0.25 in.)
fitting
1/4 in. female NPT inlet/outlet connection for pressure gauge and vent/relief valve attachment
1/4 in. female NPT inlet/outlet elbow fitting(s) for inlet and outlet (labeled)
shipped in
ambient
General description
- Closures are secured by cam-lock handle.
- Vessels are autoclavable when vented.
- Accessory code-complying relief valve, pressure gauge and hose connectors are available.
- Accessory closure adapts vessels for vacuum use.
Pressure Vessels
XX6700P01 Dispensing Pressure Vessel, 1 gal
XX6700P05 Dispensing Pressure Vessel, 5 L
XX6700P10 Dispensing Pressure Vessel, 10 L
XX6700P20 Dispensing Pressure Vessel, 20 L
Replacement Parts
6976 Vessel Cover, cam lock
6977 Outlet Dip Tube, 5 L vessel
6978 Outlet Dip Tube, 10 L vessel
6979 Outlet Dip Tube, 20 L vessel
XX6700059 O-ring, fluoroelastomer A
XX6700104 Street Elbow, 1/4 in. NPTF to M
YY1301009 Plug, 1/4 in. NPT, stainless steel
Accessories
TP0001326 PTFE Tape 12.5mm x 6.6m roll
XX6700030 Quick-release Nipple & Coupling, 1/4 in. NPTM
XX6700034 3 meters PVC tubing with 9.5 mm internal diameter
XX6700035 Tubing for pressure use, 1/2 in. ID
XX67000PK Pressure Vessel Accessory Kit
XX6700125 Hex Nipple, 1/4 in. NPTM 51 mm., stainless steel
XX6700L11 Adapter, M, 1/4 in. NPT to 1/4-1/2 in. ID
XX6700L15 Pressure Gauge, 1/4 in. ASME
XX6700L24 Vent/Relief Valve, 7 bar, ASME
XX6700L55 Bubble Point Test Apparatus
YY1301015 Pressure Gauge, 0–7 bar
YY2029348 Ball Valve, 1/4 in. NPTF, stainless steel
Application
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.
This is a User Guide Dispensing Pressure Vessel Fitting Kit from Millipore.
-
RE: dispensing pressure vessel (XX6700P01). Please provide information about weight, height and diameter of 1 Gal and 5L vessels. Connections not visible from photos. Interested in Swagelok 1/8" (or 1/4") fittings. Please provide also drawings.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service