Skip to Content
Merck
All Photos(4)

Key Documents

UFC5010

Millipore

Amicon® Ultra Centrifugal Filter, 10 kDa MWCO

sample volume 0.5 mL, regenerated cellulose membrane, MWCO 10 kDa

Synonym(s):

Centrifugal concentrator, 10 kDa MWCO, 0.5 mL sample volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

material

polypropylene cap
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing

Quality Level

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Ultra

parameter

0.5 mL sample volume

technique(s)

protein extraction: suitable
protein purification: suitable

L

4.99 cm (1.96 in.)

diam.

10.8 mm

filtration area

1 cm2

volume

0.5 mL

pore size

10 kDa MWCO

shipped in

ambient

General description

Amicon® Ultra-0.5 centrifugal filter devices provide fast ultrafiltration, with the capability for high concentration factors and easy concentrate recovery from dilute and complex sample matrices. For example, Cytochrome c (12.4 kDa) at 0.25 mg/ml initial concentration is typically retained at >95% .

The vertical design and available membrane surface area provide fast sample processing, high sample recovery (typically greater than 90% of dilute starting solution), and the capability for 30-fold concentration. Solute polarization and subsequent fouling of the membrane are minimized by the vertical design, and a physical deadstop in the filter device prevents spinning to dryness and potential sample loss. Efficient recovery of the concentrated sample (retained species) is achieved by a convenient reverse spin step after collecting the filtrate.

Amicon® Ultra-0.5 devices are supplied non-sterile and are for single use only. With their vertical membrane design and reverse spin, Amicon® Ultra 0.5 mL filters deliver great performance and lower spin times.

Application

  • Ultrafiltration with 10,000 Dalton molecular weight cutoff Ultracel® membrane
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates, primer removal, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
  • Desalting, buffer exchange, and protein dialysis

Features and Benefits

  • Ultracel® regenerated cellulose membrane with 10 kDa NMWCO delivers retentate recovery of greater than 90%
  • Concentrates 500 μL down to 15 μL
  • Concentration factor 25x to 30x
  • High sample recovery (greater than 90% of dilute starting solution)
  • Fast processing time: 10 to 30 minutes
  • Reverse spin capability provides consistent recoveries

Physical form

Color Code: Clear

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Commercially available protein labeling kits provide researchers with the flexibility to customize their own immunoassay detection panels.

Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.

PNGase Fast denaturing buffer and enzyme provide results similar to a conventional 20-hour protocol, reducing workflow time to about 1 hour.

Filtration methods and membrane filter selection in nanoparticle purification and production.

See All

Protocols

Best practices for standardizing and reducing the Extracellular Vesicle Preparation workflow, and eliminating contamination of EV preparations with dead-cell-derived vesicles.

If an antibody must be purified from a small sample volume (0.5-10 mL), then the Amicon® Pro Purification System is recommended.

Detergents are frequently used to solubilize proteins and nucleic acids during purification, but the presence of detergents may interfere with downstream analyses.

Related Content

Concentrate protein, don’t lose it. Recover protein reliably when you concentrate with Amicon® Ultra filters.

Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service