Skip to Content
Merck
All Photos(1)

Key Documents

TR-1004

Sigma-Aldrich

Cloning Cylinder, 8 mm x 8 mm diameter

Synonym(s):

lentiviral transduction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32011202
NACRES:
NA.75
Pricing and availability is not currently available.

manufacturer/tradename

Specialty Media

Quality Level

input

sample type: mouse embryonic stem cell(s)

shipped in

ambient

General description

Store at ambient temp.

Application

Research Category
Cell Culture

Packaging

15 sterile cylinders/60mm dish

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service