Skip to Content
Merck
All Photos(2)

Key Documents

SYMBFLE01

Millipore

Steritest® Symbio FLEX Pump

2 media pump kit for use in isolator or laminar flow hood

Synonym(s):

Peristaltic Pump, Sterility Testing Pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
JA.12
Pricing and availability is not currently available.

AC/DC input

100 V / 240 V, 50-60 Hz

Quality Level

material

316L stainless steel pump (Pump Head Cover, Bottle Holder, Drain Tray Support)
EPDM rubber seal
polyester keys (keypad)
polyphenylsulfone (PPSU) tubing (Tubing guide, Drain Trat Container)

Agency

EP 2.6.1
JP 4.06
USP 71

manufacturer/tradename

Steritest®

pump H

35.6 cm , with bottle holder

pump W

61.1 cm , with accessories

pump body H

18.6 cm , with display

pump body W

27.6 cm

pump body depth

36.1 cm

pump head H

8.2 cm

application(s)

pharmaceutical (Industrial Microbiology)
sterility testing

General description

The Steritest® Symbio FLEX Pump suitable for 2 media containers is uniquely designed to fit with round and oval isolator cut-outs and can be installed in multiple ways. In the standard configuration this pump is being used, integrated into an isolator, either in a low or high integration. Based on the configuration chosen, especially designed integration kits (sold separately) are offered. There are two integration kits available, one for the round cut-out and another for the oval cut-out design. However, thanks to its versatility it can also be used in a bench-top configuration for the use in a laminar flow hood, a biosafety cabinet, or in a clean room.
The automatic closure of the pump head eliminates the risk of glove pinching and ensures reliable splitting of the liquid sample. As ergonomic working conditions are key, the drain tray and bottle holder can be easily and flexibly positioned around the pump head for easy access in limited-space environments. Furthermore, the drain tray is designed to minimize splashing under the canister base. The pump′s convenient pressure detection system which can be run in two different modes (pressure monitoring and pressure regulation), reduces the risk of false test result and sterility test interruption. For highest precision and reproducible sampling also of smallest volumes, the pump has a convenient timer function available. To further ensure test reproducibility and to be always compliant with the defined test procedures, the respective test methods can be created on a PC using the Steritest® Symbio Software. These test methods will then be transferred to the pump and will lead the operator through the workflow step-by-step.
A wide range of optional accessories are also offered to improve the sterility testing workflow.

Application

Steritest® Symbio FLEX Pump (2 media) is suitable for sterility testing inside an isolator, laminar flow hood, biosafety cabinet, or cleanroom. The sterile products that can be tested with the Steritest® system include, large- and small-volume parenteral solutions in glass or plastic bottles, collapsible bags, ampoules, and vials, pre-filled syringes, lyophilized and other soluble products in ampoules or vials, antibiotics in ampoules or vials, medical devices, and difficult-to-dissolve powders.

Features and Benefits

  • Easy-to-use Design.
  • Reliable in nature.
  • Safe to use.
  • Ergonomic Design.
  • User Friendly.

Components

Steritest® Symbio FLEX pump kit is comprised of:
  • Peristaltic Pump.
  • Guides Quality Certificate.
  • Bottle Holder.
  • Drain Tray.
  • Silicone Drain Tubing.
  • Communication Hub.
  • Power Cord (Region Specific).
  • Network Cable.
  • USB Flash Drive.
  • 3 Quick Guides.
  • 2 Threaded Stems.
  • 1 Flat Seal.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

accessory

Product No.
Description
Pricing

consumable

Product No.
Description
Pricing

recommended

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service