Skip to Content
Merck
All Photos(1)

Documents

SX0550

Supelco

Sodium Fluoride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Sodium Fluoride, Disodium Fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FNa
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-667-8

Agency

meets USP/NF

Quality Level

vapor pressure

1 hPa ( 1077 °C)

Assay

≥99.5% (precipitation titration)

form

crystals

impurities

≤0.01% Insoluble matter
≤0.04% Free alkali (as NaOH)
≤0.05% Free acid (as hydrofluoric acid)
≤0.1% Sodium fluosilicate (Na2SiF6)

loss

≤0.2% loss on drying, 150°C

pH

10.2 (20 °C, 40 g/L in H2O)

mp

996 °C

solubility

soluble 42 g/L

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.01%
sulfite (SO32-): ≤0.005%

cation traces

Cu: ≤0.0002%
Fe: ≤0.002%
K: ≤0.02%
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

room temp

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (precipitative titration)99.5% minChloride (Cl)0.003% maxCopper (Cu)0.0002% maxFree acid (as hydrofluoric acid)0.05% maxFree alkali (as NaOH)0.04% maxHeavy metals (as Pb)0.001% maxInsoluble matter0.01% maxIron (Fe)0.002% maxLoss on drying (150°C)0.2% maxPotassium (K)0.02% maxSodium fluosilicate (Na2 Si F6)0.1% maxSulfate (SO4)0.01% maxSulfite (SO2)0.005% max

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service