QGARD00D3
Q-Gard® Purification Cartridge
For Milli-Q® Element / Academic / Biocel / Element / Gradient / Synthesis systems connected to DI water feed containing high silt
Synonym(s):
Q-Gard® 3 Purification Cartridge
About This Item
Recommended Products
material
resin (Mixed bed ion exchange resin)
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
Q-Gard®
parameter
1.5 L/min flow rate
technique(s)
ion chromatography: suitable
matrix
Macroporous
pore size
0.5 μm pore size (CRW5)
capacity
52 g ion exchange capacity
compatibility
for use with Milli-Q® Academic
for use with Milli-Q® Biocel
for use with Milli-Q® Element
for use with Milli-Q® Gradient
for use with Milli-Q® Synthesis
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
The Q-Gard® 3 Purification Cartridge can be used with Milli-Q® Academic / Biocel / Element / Gradient / Synthesis water purification systems.
Application
Features and Benefits
- Q-Gard® 3 Purification Cartridge is tailored to the feed-water source to optimize the performance of downstream purification media to ensure consistent results.
- Genuine water purification consumable for replacement of the Q-Gard® QGARD00D3 cartridge filter.
- Designed for trace ion removal from DI pure water containing high silt.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service