Skip to Content
Merck
All Photos(1)

Documents

PX1004

Supelco

Phosphoric Acid Meta

meets analytical specifications of USP/NF GR ACS

Synonym(s):

Metaphosphoric Acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301

Agency

meets analytical specifications of USP/NF GR ACS

Quality Level

form

crystals

contains

57-63% NaPO3 as stabilizer

concentration

33.5-36.5% (HPO3)

impurities

≤0.001% Chloride (Cl)
≤0.001% Nitrate (NO3)
≤0.005% Heavy metals (as Pb)
≤0.005% Iron (Fe)
≤0.005% Sulfate (SO4)
≤0.02% Substances reducing permanganate (as H3PO3)
≤1 ppm Arsenic (As)

color

white to clear

shipped in

ambient

storage temp.

room temp

General description

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Arsenic (As): 1ppm max
Assay (HPO3): 33.5-36.5%
Chloride (Cl): 0.001% max
Color: White or clear
Form: Crystals/lumps
Heavy metals (as Pb): 0.005% max
Iron (Fe): 0.005% max
Nitrate (NO3): 0.001% max
Stabilizer (NaPO3): 57.0-63.0%
Substances reducing permanganate (as H3PO3): 0.02% max
Sulfate (SO4): 0.005% max

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service