Skip to Content
Merck
All Photos(1)

Key Documents

PBGC07610

Millipore

PES Membrane Filter, 10 kDa NMWCO

Biomax®, filter diam. 76 mm

Synonym(s):

Biomax PES ultrafiltration membrane discs, Ultrafiltration Discs, 10 kDa NMW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

polyethersulfone membrane

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Biomax®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

diam.

76 mm

filter diam.

76 mm

filtration area

41.8 cm2

pore size

10 kDa NMWCO

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Ultrafiltration cut disc membranes in Biomax® polyethersulfone (PES) are ideal for protein concentration, purification, and desalting.
  • Optimized for concentrating or desalting serum, plasma, or conditioned tissue culture media.
  • A wide range of pore sizes is available to suit your application needs.

Application

UltrafiltrationDiscs, 10 kDa NMW are suitable for protein concentration, purification, or desalting of highly concentrated samples such as serum, plasma, or conditioned tissue culture media in bulk. It is also suitable for buffer exchange.

Features and Benefits

  • Offers discs with a wide range of pore sizes to meet your separation requirements
  • Compatible with Amicon® Stirred Cells

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service