Skip to Content
Merck

Skip To

NM0041

NovaSeal End Cover Pinch Pipes

3/8 in. ID x 5/8 in. OD - 200 pack

Synonym(s):

Silicon protection cover for crimped pinch pipes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
32010310
UNSPSC Code:
41121815
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes - NM0019PK100


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NM0039NM0022NM0021
product line

NovaSeal

product line

NovaSeal

product line

NovaSeal

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes - NM0019PK100

compatibility

for use with NovaSeal pinch pipes - NM0001PK100

compatibility

for use with NovaSeal pinch pipes - NM0019PK100

compatibility

for use with NovaSeal pinch pipes - NM0001PK100

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

Quality Level

400

Quality Level

400

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under NovaSeal End Cover Pinch Pipes




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Questions

Reviews

No rating value

Active Filters