Skip to Content
Merck

Skip To

NM0021

NovaSeal Die Kits

1/4 in. ID x 1/2 in. OD

Synonym(s):

Cutting Die for Electrical Crimping Tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32010310
UNSPSC Code:
41121815
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes - NM0001PK100


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NM0039NM0041GL105010
description

1/4 in. ID x 1/2 in. OD

description

1/4 in. ID x 1/2 in. OD - 200 pack

description

3/8 in. ID x 5/8 in. OD - 200 pack

description

-

compatibility

for use with NovaSeal pinch pipes - NM0001PK100

compatibility

for use with NovaSeal pinch pipes - NM0001PK100

compatibility

for use with NovaSeal pinch pipes - NM0019PK100

compatibility

-

Quality Level

100

Quality Level

400

Quality Level

400

Quality Level

SP1

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

-

product line

NovaSeal

product line

NovaSeal

product line

NovaSeal

product line

-


Still not finding the right product?

Explore all of our products under NovaSeal Die Kits




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters