Skip to Content
Merck

Skip To

MXPTRAY03

Milliflex® plus tray

Milliflex® Plus Pump, suitable for bioburden testing, water monitoring, stainless steel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.24
UNSPSC Code:
41116129
Manufacturer/tradename:
Milliflex®
Material:
stainless steel
Packaging:
pkg of 1 unit
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

stainless steel

Quality Segment

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex®

dilution

(Pharmaceutical product analysis)

input

sample type pharmaceutical(s)

application(s)

bioburden testing
water monitoring, pharmaceutical

compatibility

Milliflex® Plus Pump

shipped in

ambient

General description

The Manifold Tray for Milliflex® Plus Pumps allows to facilitate movement of the pumps. The tray is by default equipped with rubber feet. Another tray reference is available for use in an Isolator.

Application

The tray is an accessory of the Milliflex® Plus Pump used during filtration of the test samples for bioburden testing, and water monitoring.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MMTCNNECTMXQSEAL01MXPHEAD01
description

Milliflex® Plus Pump, suitable for bioburden testing, water monitoring, stainless steel

description

for use with Milliflex Oasis® systems, connects two pumps

description

Spare part - For use with Milliflex® Quantum system

description

-

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex Oasis®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

material

stainless steel

material

-

material

-

material

316L stainless steel

packaging

pkg of 1 unit

packaging

pack of 3 ea

packaging

-

packaging

-

Quality Level

400

Quality Level

SP1

Quality Level

SP1

Quality Level

-

application(s)

bioburden testing
water monitoring, pharmaceutical

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

application(s)

pharmaceutical

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters