Skip to Content
Merck

MXPHEAD01

Additional Milliflex pump head

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NB.79
UNSPSC Code:
41116129

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

316L stainless steel

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)
sample type bottled water
sample type beer
sample type cosmetics

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

shipped in

ambient

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MXPTEST01MMSANKITCVRMMHEADCV1
application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

application(s)

bioburden testing
pharmaceutical
sterility testing

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

ambient

material

316L stainless steel

material

-

material

stainless steel

material

-

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex Oasis®

input

sample type pharmaceutical(s)
sample type bottled water
sample type beer
sample type cosmetics

input

sample type bottled water
sample type pharmaceutical(s)
sample type beer
sample type cosmetics

input

-

input

-

General description

The compact Milliflex Plus vacuum pump streamlines filtration to accelerate QC testing. Together, the Milliflex filter units and the Milliflex Plus vacuum pump provide an integrated solution to deliver reliable and accurate results in high throughput production environments.
Sanitization is easy, so there’s minimal disruption to workflow. For even greater throughput, a high-volume workstation of up to three Milliflex PLUS pumps can be set-up and used by a single operator. Testing reveals 3 Milliflex PLUS pumps are faster than a 6-place manifold.

Preparation Note

Sterilization Method
Autoclavable

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Milliflex® Quantum Rapid Detection System An easy-to-use, non-destructive, fluorescent staining-based system for faster microbial detection

See All

Questions

  1. What is the flow rate, max vacuum, and pressure of this system?

    1 answer
    1. As per the user guide - Flow rate: < 16 sec/ml for 100 ml of MilliQ water filtered through MXHA membrane. Accuracy: +/- 5% of 100 ml

      Most of issues related to flow rate are due to incorrect weight calibration.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service