Skip to Content
Merck
All Photos(2)

Key Documents

MMHEADQU1

Millipore

MILLIFLEX® Quantum membrane transfer tool

for use with Milliflex Oasis® System for Milliflex® Quantum application

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79
Pricing and availability is not currently available.

material

316L stainless steel
stainless steel head

Quality Level

packaging

pkg of 1 unit (per pack)

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® System for Milliflex® Quantum application

shipped in

ambient

Related Categories

General description

Milliflex® Quantum membrane transfer tool is an additional accessory for using Milliflex® Quantum Rapid Detection System with the Milliflex Oasis® system. Milliflex® Quantum System is a rapid fluorescence-based technology for fast quantitative detection of contaminants over a broad range of filterable matrices. This easy to use and simple system can detect viable microorganisms filtered on a membrane down to one CFU per sample. The Milliflex® Quantum system uses proven technology based on a universal enzymatic fluorescent staining of viable microorganisms. The Milliflex® Quantum membrane transfer tool should be used in a microbiological safety cabinet or laminar flow hood. Place the Milliflex® Quantum membrane transfer tool onto the pump and lock it by turning clockwise until the mark on the head is aligned with the one on the pump body.

Application

Milliflex® Quantum membrane transfer tool is an additional accessory for using Milliflex® Quantum Rapid Detection System with Milliflex Oasis® system. It ensures safety removal of the membrane. It finds application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical products analysis.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service