Skip to Content
Merck

Skip To

MXQSUP001

Milliflex® Quantum Reader Stand

For use with Milliflex® Quantum system

Synonym(s):

Stainless steel stand

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79
eCl@ss:
32160606
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

304L stainless steel

Quality Segment

manufacturer/tradename

Milliflex®

application(s)

pharmaceutical

shipped in

ambient

General description

Milliflex® Quantum Reader Stand is an additional accessory for Milliflex® Quantum Rapid Detection System. Milliflex® Quantum System is a rapid fluorescence-based technology for fast quantitative detection of contaminants over a broad range of filterable matrices proving as a fast and reliable alternative for the detection of microorganisms. The Reader stand consists of adjustment knobs for the firm positioning of the reader. The stand is made of stainless steel with Styrenebutadiene rubber foot and silicone end stops aiding a stable support system.

Application

Milliflex® Quantum Reader stand is an additional accessory for the Milliflex® Quantum Rapid Detection System to ensure firm positioning of the reader. It finds application in bioburden testing and water monitoring in Pharmaceuticals industry.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MXQREAD01MMQUANK01MXQTRT001
description

For use with Milliflex® Quantum system

description

Part of the Milliflex® Quantum system, Fluorescence-based technology, For the rapid detection of microbial contamination

description

Supplied with 1 Reader, 1 Reader Stand, 1 Membrane Transfer Tool, 1 Removal Rack, 1 Camera and Installation flash drive

description

For use with Milliflex® Quantum system

shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

ambient

material

304L stainless steel

material

304L stainless steel chamber (optical), 304L stainless steel handle, PVC foot, aluminum housing (sheet 1050), glass filter (OG550; optical), polyester housing, transparent polycarbonate protector(s) (LED)

material

304L stainless steel (Camera front plate), 304L stainless steel (Optical Chamber, Reader), 304L stainless steel handle (Reader), 304L stainless steel stand (Reader), PVC foot (Reader), aluminum foot (with epoxy coating, Camera), aluminum housing (Sheet, Reader), glass window(s) (Camera), polyoxymethylene housing (Membrane Transfer Tool), polyphenylsulfone (PPSU) (Removal Rack Membrane Transfer Tool ), silicone seal (Membrane Transfer Tool)

material

black polyoxymethylene housing, silicone seal, stainless steel body (1.4462), stainless steel lever(s) (1.4462)

Quality Level

SP1

Quality Level

EQ2

Quality Level

EQ2

Quality Level

SP2

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters