Select a Size
About This Item
Skip To
AC/DC input
100 V / 240 V AC, 50-60 Hz (Auto-detectable (PFC compliant))
description
configured for Water, Isopropyl Alcohol (95%), Ethanol (95%), Dimethyl - sulfoxide (70%), Methanol (95%), Acetonitrile (75%), Bleach (70%), Xylene (95%), Acetone (95%)
parameter
-196-100 °C temp. range
compatibility
configured for Water, Isopropyl Alcohol (95%), Ethanol (95%), Dimethyl - sulfoxide (70%), Methanol (95%), Acetonitrile (75%), Bleach (70%), Xylene (95%), Acetone (95%), for use with MilliSentials Lab Adaptable Labels
Quality Level
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | |||
|---|---|---|---|
| AC/DC input 100 V / 240 V AC, 50-60 Hz (Auto-detectable (PFC compliant)) | AC/DC input - | AC/DC input - | AC/DC input - |
| description configured for Water, Isopropyl Alcohol (95%), Ethanol (95%), Dimethyl - sulfoxide (70%), Methanol (95%), Acetonitrile (75%), Bleach (70%), Xylene (95%), Acetone (95%) | description - | description - | description - |
| parameter -196-100 °C temp. range | parameter - | parameter - | parameter - |
| compatibility configured for Water, Isopropyl Alcohol (95%), Ethanol (95%), Dimethyl - sulfoxide (70%), Methanol (95%), Acetonitrile (75%), Bleach (70%), Xylene (95%), Acetone (95%) | compatibility - | compatibility - | compatibility - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
General description
Properties-
1. MilliSentials Lab Printer-
- Width:202 mm (8.0 in)
- Height:192 mm (7.5 in)
- Weight:2.5 kg (5.5 lbs)
- Connection / Communication: USB, F802.11ac WIFI, Bluethooth 4.1
- Resolution:300 dpi
- Power:Auto-detectable (PFC compliant), 100-240 VAC, 50-60Hz
- Warranty: 3 year
2. MilliSential Lab Adaptable Labels-
- Label: MilliSentials Lab Adaptable Labels
- Vessel Compatibility:0.5 mL Tubes, 0.6 mL Tubes, HPLC vials, 1 mL Tubes, 2 mL Tubes, Cryovials, Plates, Petri Dishes, 15 mL Conical Tubes, 50 mL Conical Tubes, Any Vessel Larger than 5 mL
- Temperature Range:-196°C to 100°C
- Chemical Compatibility: Water, Isopropyl Alcohol (≤95%), Ethanol (≤95%), Dimethyl - sulfoxide (≤70%), Methanol (≤95%), Acetonitrile (≤75%), Bleach (≤70%), Xylene (≤95%), Acetone (≤95%)
3. MilliSentials Lab Printer Ribbon-
Ribbon Capacity: ∼6400 labels printed from 1 roll of ribbon
4. MilliSentials Lab Labeling System Software - USB Key
- Software: MilliSentials Lab Labeling System Software
- Software Compatibility: Microsoft Windows 10 OS, version 1909 OS Build 18363.1016 Macintosh(Mac) macOS Catalina, version 10.15.6
Application
Other Notes
2. MilliSential Lab Adaptable Labels -1 roll
3. MilliSentials Lab Printer Ribbon - 1 roll
4. MilliSentials Lab Labeling System Software - USB Key
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instrumentos de laboratorio intuitivos, herramientas ergonómicas, datos accesibles y soluciones con visión de futuro para laboratorios de investigación, biotecnología y farmacéuticos.
Agilice la identificación de muestras, reactivos y productos químicos utilizando el sistema de etiquetado de laboratorio MilliSentials™: etiquetas impermeables y adaptables a la medida de los laboratorios de investigación.
MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.
Streamlined, ergonomic, essential lab equipment solutions for handling, labeling, pipetting, aliquoting, and homogenizing samples in research laboratories.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service