Skip to Content
Merck

Skip To

MISELLABSI

MilliSentials Lab Labeling System

India

Synonym(s):

Laboratory Labeling System

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
44102408
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

General description

The MilliSentials Lab Labeling System provides a complete laboratory labeling solution with laboratory grade labels, a compact WIFI capable printer, and custom-designed laboratory labeling software. The laboratory labels are capable of withstanding temperatures from -196°C (liquid nitrogen) to 100°C (boiling water) and provide resistance to harsh chemical solvents (including ethanol, DMSO, xylene). The labels have also been designed with perforations allowing one size to fit across a wide range of laboratory vessels including microcentrifuge tubes, cryovials, petri dishes, cell culture plates, and reagent bottles. The custom-built software features pre-constructed label templates for laboratory vessels, allows labels to be saved for quick repeat printing, and features a spreadsheet upload function for large batch printing. The MilliSentials Lab Labeling System offers a complete labeling system specifically designed for the laboratory.

Properties-
1. MilliSentials Lab Printer-
Width:202 mm (8.0 in)
Height:192 mm (7.5 in)
Weight:2.5 kg (5.5 lbs)
Connection / Communication: USB, F802.11ac WIFI, Bluethooth 4.1
Resolution:300 dpi
Power:Auto-detectable (PFC compliant), 100-240 VAC, 50-60Hz
Warranty: 3 year

2. MilliSential Lab Adaptable Labels-
Label: MilliSentials Lab Adaptable Labels
Vessel Compatibility:0.5 mL Tubes, 0.6 mL Tubes, HPLC vials, 1 mL Tubes, 2 mL Tubes, Cryovials, Plates, Petri Dishes, 15 mL Conical Tubes, 50 mL Conical Tubes, Any Vessel Larger than 5 mL
Temperature Range:-196°C to 100°C
Chemical Compatibility: Water, Isopropyl Alcohol (≤95%), Ethanol (≤95%), Dimethyl - sulfoxide (≤70%), Methanol (≤95%), Acetonitrile (≤75%), Bleach (≤70%), Xylene (≤95%), Acetone (≤95%)

3. MilliSentials Lab Printer Ribbon-
Ribbon Capacity: ∼6400 labels printed from 1 roll of ribbon

4. MilliSentials Lab Labeling System Software - USB Key
Software: MilliSentials Lab Labeling System Software
Software Compatibility: Microsoft Windows 10 OS, version 1909 OS Build 18363.1016 Macintosh(Mac) macOS Catalina, version 10.15.6

Application

Laboratory Sample Labeling

Other Notes

1. MilliSentials Lab Printer
2. MilliSential Lab Adaptable Labels -1 roll
3. MilliSentials Lab Printer Ribbon - 1 roll
4. MilliSentials Lab Labeling System Software - USB Key

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MISELLABSEMISELLABSLMISELLABSN
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under MilliSentials Lab Labeling System




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Agilice la identificación de muestras, reactivos y productos químicos utilizando el sistema de etiquetado de laboratorio MilliSentials™: etiquetas impermeables y adaptables a la medida de los laboratorios de investigación.

Instrumentos de laboratorio intuitivos, herramientas ergonómicas, datos accesibles y soluciones con visión de futuro para laboratorios de investigación, biotecnología y farmacéuticos.

MilliSentials™ Lab Labeling System offers concise, temperature-resistant labels, simplifying laboratory labeling.

View All Related Content




Questions

Reviews

No rating value

Active Filters