MAFBN0B50
Multiscreen GFB Filter Plate, Opaque Non-sterile
Receptor/ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification
About This Item
Recommended Products
material
Barex®
PVDF membrane (hydrophilic)
flat bottom wells
glass fiber membrane
titanium dioxide
Quality Level
sterility
non-sterile
product line
MultiScreen®
feature
hydrophilic
lid
manufacturer/tradename
MultiScreen®
parameter
50-250 μL sample volume (per well)
technique(s)
DNA purification: suitable
activity assay: suitable (with precipitations)
ligand binding assay: suitable
receptor binding assay: suitable
H
14.4 mm
L
128 mm
W
85.5 mm
filtration area
0.26 cm2
plate size
96 wells
well maximum volume
300 μL
working volume
50-250 μL
pore size
1.0 μm pore size
application(s)
sample preparation
binding type
high binding surface
shipped in
ambient
Related Categories
General description
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particular laden samples. They are also widely used for clarification of aqueous solutions.
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particulate-laden samples. They are also widely used for clarification of aqueous solutions.
Features & Benefits:
- Multiple configurations and applications
- Chemical compatibility
- Automation-compatible
- Superior vacuum filtration and filtrate collection
Applications:
- Receptor/Ligand Binding Assays, Sample Preparation, TCA Precipitation of Protein
Application
General Filtration
Nucleic acid
Assays
Physical form
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
MultiScreenHTS 384-well filter plate for receptor-ligand binding assays using Human muscarinic M1 GPCR.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service