Skip to Content
Merck
All Photos(1)

Key Documents

MABT827-AF555

Sigma-Aldrich

Anti-Dystrophin Antibody, clone 2C6 (MANDYS106), Alexa Fluor 555 Conjugate

clone 2C6 (MANDYS106), from mouse, ALEXA FLUOR 555

Synonym(s):

Dystrophin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
conjugate:
ALEXA FLUOR 555
application:
IF
clone:
2C6 (MANDYS106), monoclonal
species reactivity:
human
citations:
technique(s):
immunofluorescence: suitable
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C6 (MANDYS106), monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DMD(1756)

General description

Dystrophin (UniProt P11532) is encoded by the DMD (also known as BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85) gene (Gene ID 1756) in human. Dystrophin is localized to the inner part of the muscle fiber cell membrane (sarcolemma), where it forms the dystrophin-associated glycoprotein complex (DGC) that links the extracellular matrix to the actin cytoskeleton. Dystrophin plays an important role in stabilizing the muscle fibre against the mechanical forces of muscle contraction by providing a shock-absorbing connection between the cytoskeleton and the extracellular matrix. Duchenne muscular dystrophy (DMD) is caused by gene mutations that disrupt the open reading frame (ORF) and prevent the full translation of dystrophin. ORF restoration by exon skipping using antisense oligonucleotides targeted to splicing elements are designed to transform the DMD phenotype to that of the milder disorder, Becker muscular dystrophy (BMD), typically caused by in-frame dystrophin deletions that allow the production of an internally deleted but partially functional dystrophin.

Application

The unconjugated antibody (Cat. No. MABT827) and Alexa Fluor 488 conjugate (Cat. No. MABT827-AF488) are also available for Immunohistochemistry, Immunofluorescence, and Western Blotting applications.
This mouse monoclonal Anti-Dystrophin Antibody, clone 2C6 (MANDYS106), Alexa Fluor 555 Conjugate, Cat. No. MABT827-AF555 is validated for use in Immunofluorescence for the detection of Dystrophin.

Quality

Evaluated by Immunofluorescence in human skeletal muscle tissue sections.

Immunofluorescence Analysis: A 1:100 dilution of this antibody detected dystrophin in human skeletal muscle tissue sections.

Target description

~427 kDa observed.

Physical form

Purified mouse IgG2a Alexa Fluor 555 conjugate in buffer containing PBS, 1.5% BSA with 0.05% sodium azide

Analysis Note

Control
The unconjugated antibody (Cat. No. MABT827) and Alexa Fluor 488 conjugate (Cat. No. MABT827-AF488) are also available for Immunohistochemistry

Immunofluorescence

and Western Blotting applications.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service