Skip to Content
Merck
All Photos(1)

Documents

MABS15

Sigma-Aldrich

Anti-IRS-2 Antibody, clone 9.5.2

clone 9.5.2, from mouse

Synonym(s):

insulin receptor substrate 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9.5.2, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IRS2(8660)

General description

Insulin receptor substrate (IRS) molecules are key mediators in insulin signaling and play a central role in maintaining basic cellular functions such as growth, survival, and metabolism. They act as docking proteins between the insulin receptor and a complex network of intracellular signaling molecules containing Src homology 2 (SH2) domains. Four members (IRS-1, IRS-2, IRS-3, IRS-4) of this family have been identified that differ as to tissue distribution, subcellular localization, developmental expression, binding to the insulin receptor, and interaction with SH2 domain-containing proteins. In fact, IRS-1 appears to have its major role in skeletal muscle whereas IRS-2 appears to regulate hepatic insulin action as well as pancreatic beta cell development and survival. By contrast, IRS-3 and IRS-4 genes appear to play a redundant role in the IRS signaling system. Defects in muscle IRS-1 expression and function have been reported in insulin-resistant states such as obesity and type 2 diabetes.

Specificity

This antibody recognizes IRS-2.

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to mouse IRS-2.

Application

Anti-IRS-2 Antibody, clone 9.5.2 detects level of IRS-2 & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

Evaluated by Western Blot in Insulin treated CHO IR/IRS-2 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected IRS-2 on 10 µg of Insulin treated CHO IR/IRS-2 cell lysate.

Target description

~ 185 kDa

Linkage

Replaces: 06-506

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Insulin treated CHO IR/IRS-2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Kuznetsova et al.
JCI insight, 1(3) (2016-05-07)
The capacity of pancreatic β cells to maintain glucose homeostasis during chronic physiologic and immunologic stress is important for cellular and metabolic homeostasis. Insulin receptor substrate 2 (IRS2) is a regulated adapter protein that links the insulin and IGF1 receptors
Gerta Hoxhaj et al.
PloS one, 8(9), e73327-e73327 (2013-09-17)
Insulin receptor substrate 1 (IRS1) and IRS2 are well-characterized adapter proteins that relay signals from receptor tyrosine kinases to downstream components of signalling pathways. In contrast, the function of IRS4 is not well understood. IRS4 overexpression has been associated with
A Mora et al.
Journal of immunology (Baltimore, Md. : 1950), 166(4), 2218-2227 (2001-02-13)
Proliferative responses of lymphoid cells to IL-2 and IL-4 depend on activation of the cells, but the mechanism(s) by which activation enhances cellular competence to respond to cytokines is not fully understood. The NF-kappaB/Rel family represents one signal transduction pathway
Nipun Saini et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 37(10), e23172-e23172 (2023-09-04)
Prenatal alcohol exposure (PAE) impairs fetal growth and neurodevelopment. Although alcohol is well known to alter metabolism, its impact on these processes during pregnancy is largely unexplored. Here, we investigate how alcohol affects maternal-fetal glucose metabolism using our established mouse
Zheng Sun et al.
BMC complementary and alternative medicine, 16, 200-200 (2016-07-10)
Disturbance in energy metabolism, as a key factor in diabetes-associated cognitive decline (DACD), has become a promising therapeutic target of Chinese medicine ZiBu PiYin Recipe (ZBPYR). However, it is still not clear how ZBPYR affects the mitochondrial function in DACD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service