Skip to Content
Merck
All Photos(1)

Key Documents

804614

Sigma-Aldrich

Bovine Collagen Solution

Type I, Acid soluble telocollagen, 6 mg/mL, sterile filtered, BSE-Free, suitable for biomedical research

Synonym(s):

Collagen I

Sign Into View Organizational & Contract Pricing

Select a Size

20 ML
$904.00

$904.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 ML
$904.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$904.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

bovine (collagen)

form

liquid

concentration

6 mg/mL

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

  • Bovine corium, acid solubilized collagen, telocollagen, pH 2 in 0.01 N HCl. Predominantly Type I collagen with approximately 5% Type III, at 6 mg/ml.
  • Acid extracted collagen with telopeptide-intact.
  • Sterile filtered.
  • BSE-Free.

Application

Forms a gel and is ideal for 3D biomaterial scaffolds for cell culture, engineering tissue,[1] and drug delivery.[2][3]

Legal Information

Product of Collagen Solutions.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wallace D, et al.
Advanced Drug Delivery Reviews, 55, 1631-1649 (2013)
Bareil RP, et al.
Materials, 3, 1863-1887 (2010)
Luo Z, et al.
Angewandte Chemie (International Edition in English), 50, 640-643 (2011)

Articles

Collagen-based delivery systems effectively deliver a range of viral and non-viral gene carriers by providing spatiotemporal control over delivery and reducing degradation rates and immune responses.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

See All

Questions

  1. What is the different between this product (#804614) and #804622?

    1 answer
    1. The only differences between the two types of Bovine Collagen Solution is the testing performed on the materials. Please view the sample documentation for more information on both products.

      Product 804614:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/804614-20MLSLCP3797.pdf
      https://www.sigmaaldrich.com/coo/undefined/804614/*

      Product 804622:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/804622-20MLSLCM9858.pdf
      https://www.sigmaaldrich.com/coo/undefined/804622/*

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service