Skip to Content
Merck
All Photos(2)

Key Documents

MABE656

Sigma-Aldrich

Anti-HP1γ Antibody, clone 14D3.1

clone 14D3.1, from mouse

Synonym(s):

Chromobox protein homolog 3, HECH, Heterochromatin protein 1 homolog gamma, HP1 gamma, Modifier 2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

14D3.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology), nonhuman primates (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CBX3(11335)

General description

Chromobox protein homolog 3 (UniProt Q13185; also known as HECH, Heterochromatin protein 1 homolog gamma, HP1 gamma, Modifier 2 protein) is encoded by the CBX3 gene (Gene ID 11335) in human. HP1 is a non-histone chromosomal protein that is present as three orthologs, HP1-alpha, HP1-beta, and HP1-gamma, in mammals. While HP1-alpha and HP1-beta have been unambiguously localized in heterochromatin, HP1-gamma has been found to localize in both euchromatin and heterochromatin (transiently) compartments of interphase nuclei, as well as in the pericentromeric chromatin and arms of mitotic chromosomes of 3T3 cells. HP1-gamma is phosphorylated by PIM1 during interphase and may be hyper-phosphorylated during mitosis. HPI-gamma is involved in transcriptional silencing in heterochromatin-like complexes. It recognizes and binds histone H3 tails methylated at ′Lys-9′, leading to epigenetic repression. It may also contribute to the association of the heterochromatin with the inner nuclear membrane through its interaction with lamin B receptor (LBR). It is also reported to be involved in the formation of functional kinetochore through interaction with MIS12 complex proteins. Ref.: Minc E., et al. (2000). Cytogenet. Cell Genet. 90, 279-284.
Lachner, M., et al. (2001). Nature 410, 116-120.

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human HP1γ.

Application

Anti-HP1γ, clone 14D3.1, Cat. No. MABE656, is a highly specific mouse monoclonal antibody that targets heterochromatin protein-1 gamma and has been tested in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected nuclear HP1γ immunoreactivity in 4% paraformaldehyde-fixed, 0.1 Triton X-100-permeabilized A431 and HeLa cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected HP1γ in 10 µg of HeLa cell lysate.

Target description

~20 kDa observed. 20.68 kDa(Met1 removed) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: MAB3450

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaohua Hu et al.
The EMBO journal, 39(12), e104133-e104133 (2020-04-30)
Long non-coding RNAs (lncRNAs) are emerging regulators of genomic stability and human disease. However, the molecular mechanisms by which nuclear lncRNAs directly contribute to DNA damage responses remain largely unknown. Using RNA antisense purification coupled with quantitative mass spectrometry (RAP-qMS)
Yihan Peng et al.
Nature communications, 10(1), 1224-1224 (2019-03-16)
Poly-(ADP-ribose) polymerase inhibitors (PARPi) selectively kill breast and ovarian cancers with defects in homologous recombination (HR) caused by BRCA1/2 mutations. There is also clinical evidence for the utility of PARPi in breast and ovarian cancers without BRCA mutations, but the
Ali Fathi et al.
Aging cell, 21(1), e13541-e13541 (2021-12-26)
Modeling age-related neurodegenerative disorders with human stem cells are difficult due to the embryonic nature of stem cell-derived neurons. We developed a chemical cocktail to induce senescence of iPSC-derived neurons to address this challenge. We first screened small molecules that
Kosuke Sako et al.
The Journal of cell biology, 223(9) (2024-05-23)
Maintenance of ploidy depends on the mitotic kinase Aurora B, the catalytic subunit of the chromosomal passenger complex (CPC) whose proficient activity is supported by HP1 enriched at inner centromeres. HP1 is known to associate with INCENP of the CPC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service