Merck
  • The deubiquitylating enzyme USP15 regulates homologous recombination repair and cancer cell response to PARP inhibitors.
The deubiquitylating enzyme USP15 regulates homologous recombination repair and cancer cell response to PARP inhibitors.
Nature communications (2019-03-16)
Yihan Peng, Qingchao Liao, Wei Tan, Changmin Peng, Zhaohua Hu, Yali Chen, Zhuqing Li, Jing Li, Bei Zhen, Wenge Zhu, Xiangpan Li, Yi Yao, Qibin Song, Chengsheng Liu, Xiangdong Qi, Fuchu He, Huadong Pei
ABSTRACT
Poly-(ADP-ribose) polymerase inhibitors (PARPi) selectively kill breast and ovarian cancers with defects in homologous recombination (HR) caused by BRCA1/2 mutations. There is also clinical evidence for the utility of PARPi in breast and ovarian cancers without BRCA mutations, but the underlying mechanism is not clear. Here, we report that the deubiquitylating enzyme USP15 affects cancer cell response to PARPi by regulating HR. Mechanistically, USP15 is recruited to DNA double-strand breaks (DSBs) by MDC1, which requires the FHA domain of MDC1 and phosphorylated Ser678 of USP15. Subsequently, USP15 deubiquitinates BARD1 BRCT domain, and promotes BARD1-HP1γ interaction, resulting in BRCA1/BARD1 retention at DSBs. USP15 knockout mice exhibit genomic instability in vivo. Furthermore, cancer-associated USP15 mutations, with decreased USP15-BARD1 interaction, increases PARP inhibitor sensitivity in cancer cells. Thus, our results identify a novel regulator of HR, which is a potential biomarker for therapeutic treatment using PARP inhibitors in cancers.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Ubiquitinylated proteins Antibody, clone FK2, clone FK2, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-HP1γ Antibody, clone 14D3.1, clone 14D3.1, from mouse
Sigma-Aldrich
Anti-MDC1 Antibody, clone P2B11, clone P2B11, from mouse