MAB970
Anti-Mycoplasma bovis Antibody, Strain M23, clone MYB163
ascites fluid, clone MYB163, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
ascites fluid
clone
MYB163, monoclonal
species reactivity
bovine
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1κ
shipped in
wet ice
Specificity
Clone MYB163 reacts with multiple strains of Mycoplasma bovis. However, it does not react with any other species of Mycoplasma tested.
Immunogen
Epitope: Strain M23
Mycoplasma bovis strain M23.
Application
Anti-Mycoplasma bovis Antibody, Strain M23, clone MYB163 detects level of Mycoplasma bovis & has been published & validated for use in ELISA, WB, IH, IH(P).
ELISA at 1:2000,
Western Blot 1:200,
Immunohistochemistry 1:2000
Immunohistochemistry in formalin fixed paraffin tissues: {see Adegboye DS, et al J Vet. Diagn Invest 7(3):333-337, 1995}.
Final working dilutions must be determined by end user.
Western Blot 1:200,
Immunohistochemistry 1:2000
Immunohistochemistry in formalin fixed paraffin tissues: {see Adegboye DS, et al J Vet. Diagn Invest 7(3):333-337, 1995}.
Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Bacterial
Infectious Diseases - Bacterial
Physical form
Ascites fluid contain 10% buffered formalin.
Storage and Stability
Maintain at -20°C for up to 12 months. For short-term storage (0-4 weeks), maintain at +2-8°C Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Carlos A Margineda et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 34(4), 693-698 (2022-05-21)
HoBi-like pestivirus (HoBiPeV) is an emerging virus that has been detected in cattle and other ruminants. We diagnosed 2 cases of fatal bovine respiratory disease complex (BRDC) associated with infection with HoBiPeV in a feedlot in Argentina. The main findings
Mycoplasma bovis real-time polymerase chain reaction assay validation and diagnostic performance.
Kristin A Clothier,Dianna M Jordan,Curtis J Thompson,Joann M Kinyon,Timothy S Frana,Erin L Strait
Journal of Veterinary Diagnostic Investigation : Official Publication of the American Association of Veterinary Laboratory Diagnosticians, Inc null
Katarzyna Dudek et al.
BMC veterinary research, 15(1), 371-371 (2019-10-28)
Mycoplasma bovis is a causative agent of disease in cattle causing many clinical conditions. Currently there are no commercial M. bovis vaccines in Europe and treatment is difficult with decreased antimicrobial susceptibility of M. bovis field isolates. Using an M.
Germán Cantón et al.
Revista Argentina de microbiologia (2022-05-24)
Bovine respiratory disease (BRD) is one of the most frequent clinical concerns in weaned calves after their arrival at the feedlot. This work reports the first local isolation of Mycoplasma bovis from feedlot calves with pneumonia and polyarthritis in Argentina.
Innate immune response to intramammary Mycoplasma bovis infection.
A C W Kauf,R F Rosenbusch,M J Paape,D D Bannerman
Journal of Dairy Science null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service