Skip to Content
Merck
All Photos(1)

Key Documents

MAB80125

Sigma-Aldrich

Anti-Human Metapneumovirus Antibody, clone 134

clone 134, Chemicon®, from mouse

Synonym(s):

hMPV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

clone

134, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

General description

Human metapneumovirus (hMPV) is a member of the pneumovirinae subfamily of paramyxoviruses, first described in 2001 from pediatric respiratory specimens in the Netherlands. It is an enveloped pleomorphic virus with a single negative-strand RNA genome. Two major serotypes, A and B, have been described. Several studies identified hMPV in specimens worldwide, and estimated that by age 5 years 70% of children have developed antibodies to hMPV.

Specificity

Specific to the fusion protein. Reactive with all sub-types; A1, A2, B1, B2.

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Human Metapneumovirus Antibody, clone 134 is validated for use in ELISA, FC, IF for the detection of Human Metapneumovirus.

Physical form

Format: Purified
Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide.

Storage and Stability

Store at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service