Skip to Content
Merck
All Photos(1)

Key Documents

890880C

Avanti

18:0 TAP

1,2-stearoyl-3-trimethylammonium-propane (chloride salt), chloroform

Synonym(s):

1,2-stearoyl-3-trimethylammonium-propane (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$284.00
200 MG
$591.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$284.00
200 MG
$591.00

About This Item

Empirical Formula (Hill Notation):
C42H84NO4Cl
CAS Number:
Molecular Weight:
702.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (890880C-25mg)
pkg of 2 × 4 mL (890880C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890880C

concentration

10 mg/mL (890880C-25mg)
25 mg/mL (890880C-200mg)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Cl-]

InChI

1S/C42H84NO4.ClH/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-41(44)46-39-40(38-43(3,4)5)47-42(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;/h40H,6-39H2,1-5H3;1H/q+1;/p-1

InChI key

NTTZBBIBMSBLNK-UHFFFAOYSA-M

Application

18:0 TAP has been used in chloroform to prepare microbubbles.[1]

Packaging

5 mL Clear Glass Sealed Ampule (890880C-200mg)
5 mL Clear Glass Sealed Ampule (890880C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo transfection and detection of gene expression of stem cells preloaded with DNA-carrying microbubbles
Tavri S, et al.
Radiology, 276(2), 518-525 (2015)
A E Regelin et al.
Biochimica et biophysica acta, 1464(1), 151-164 (2000-03-08)
In order to investigate the relationship between lipid structure and liposome-mediated gene transfer, we have studied biophysical parameters and transfection properties of monocationic DOTAP analogs, systematically modified in their non-polar hydrocarbon chains. Stability, size and (by means of anisotropy profiles)
Yuki Takechi-Haraya et al.
Langmuir : the ACS journal of surfaces and colloids, 34(26), 7805-7812 (2018-06-06)
It has recently been recognized that the mechanical properties of lipid nanoparticles play an important role during in vitro and in vivo behaviors such as cellular uptake, blood circulation, and biodistribution. However, there have been no quantitative investigations of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service