Skip to Content
Merck
All Photos(1)

Documents

MAB316

Sigma-Aldrich

Anti-GABA Antibody, clone 5A9

clone 5A9, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5A9, monoclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GABRA1(2554)

Specificity

Specific for GABA and does not cross-react with other amino acids.

Immunogen

GABA-coupled to BSA with glutaraldehyde.

Application

Anti-GABA Antibody, clone 5A9 detects level of GABA & has been published & validated for use in ELISA, IH & WB.
Immunohistochemistry on frozen sections if perfusion fixed. Glutaraldehyde must be included in fixative with paraffin sections: 1:200-1:400. Typical fixative might be 4% PFA, 1% glutaraldehyde in 0.1M sodium phosphate buffer, pH 7.3 .

Immunoblotting not recommended due to low molecular wgt: 103Da.

Optimal working dilution must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal in PBS, pH 7.4, 1% BSA, and 0.1% sodium azide as a preservative.

Storage and Stability

Maintain refrigerated at 2–8°C for up to 6 months.

Analysis Note

Control
Myeloblasts, lymphocytes, erythroblasts

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shogo Soma et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(3), 485-502 (2018-11-28)
It is well known that the posterior parietal cortex (PPC) and frontal motor cortices in primates preferentially control voluntary movements of contralateral limbs. The PPC of rats has been defined based on patterns of thalamic and cortical connectivity. The anatomical
Masakazu Uematsu et al.
Cerebral cortex (New York, N.Y. : 1991), 18(2), 315-330 (2007-05-23)
Although neocortical GABAergic (gamma-aminobutyric acidergic) interneurons have been the focus of intense study, especially in the rat, a consensus view of the functional diversity and organization of inhibitory cortical neurons has not yet been achieved. To better analyze GABAergic neurons
Neelima Bhargava et al.
In vitro cellular & developmental biology. Animal, 46(8), 685-692 (2010-07-16)
A very small population of choline acetyltransferase (ChAT) immunoreactive cells is observed in all layers of the adult hippocampus. This is the intrinsic source of the hippocampal cholinergic innervation, in addition to the well-established septo-hippocampal cholinergic projection. This study aimed
Naomi Karoubi et al.
Frontiers in neuroanatomy, 10, 106-106 (2016-11-29)
Over recent years, the seven-spot archerfish (Toxotes chatareus) has emerged as a new model for studies in visual and behavioral neuroscience thanks to its unique hunting strategy. Its natural ability to spit at insects outside of water can be used
Alain C Burette et al.
The Journal of comparative neurology, 522(9), 2164-2178 (2014-03-19)
The 53 kDa insulin receptor substrate protein (IRSp53) is highly enriched in the brain. Despite evidence that links mutations of IRSp53 with autism and other neuropsychiatric problems, the functional significance of this protein remains unclear. We used light and electron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service