Skip to Content
Merck

Skip To

IM40

Anti-MMP-7 (Ab-1) Mouse mAb (141-7B2)

liquid, clone 141-7B2, Calbiochem®

Synonym(s):

Anti-Matrilysin, Anti-Matrix Metalloproteinase 7, Anti-PUMP-1

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
141-7B2, monoclonal
Species reactivity:
human
Application:
Citations:
-


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

141-7B2, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (5-10 µg/mL,
Paraffin Sections )

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP7(4316)

General description

Purified mouse monoclonal antibody. Recognizes the ~28 kDa latent form of MMP-7.
Recognizes the ~28 kDa latent form of MMP-7 in TPA-treated SW620 cells. Does not recognize the active form of MMP-7.
This Anti-MMP-7 (Ab-1) Mouse mAb (141-7B2) is validated for use in Immunoblotting, Paraffin Sections for the detection of MMP-7 (Ab-1).

Immunogen

Human
human rectal carcinoma MMP-7

Application


Immunoblotting (5-10 g/ml, see application references)
Paraffin Sections (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 100 mM sodium phosphate buffer, 0.1% BSA, pH 7.0.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
CaR-1 or TPA treated SW620 cells

Other Notes

Brunner, K.L, et al. 1995. Proc. Natl. Acad. Sci. USA92, 7362.
Imai, K., et al. 1995. J. Biol. Chem.270, 6691.
Lichtinghagen, R., et al. 1995. Eur. J. Clin. Chem. Clin. Biochem.33, 65.
Nakano, A., et al. 1995. J. Neurosurg.83, 298.
Takino, T., et al. 1995. J. Biol. Chem.270, 23013.
Woessner, J.F. 1991. FASEB J.5, 2145.
Yamamoto, H., et al. 1995. J. Clin. Lab. Anal.9, 297.
Gaire, M., et al. 1994. J. Biol. Chem.269, 2032.
McDonnell, S., et al. 1994. Biochem. Soc. Trans.22, 58.
Cottam, D.W. and Rees, R.C. 1993. Intl J. Oncol.2, 861.
Stetler-Stevenson, W.G., et al. 1993. FASEB J.7, 1434.
Woessner, J.F. 1991. FASEB J.5, 2145.
Liotta, L.A. and Stetler-Stevenson, W.G. 1990. in Sem. Cancer Biol., ed. M. M. Gottesman. Vol. 1(2), 99.
Woessner, J.F., Jr and Taplin, C.J. 1988. J. Biol. Chem.263, 16918.
Sellers, A. and Woessner, J.F., Jr. 1980. Biochem. J.189, 521.
Does not recognize the ~19 kDa active form of MMP-7. To detect active MMP-7, use Cat. No. IM47L. Does not cross-react with human MMP-1, MMP-2, MMP-3, MMP-8, MMP-9 or MMP-13. For optimal detection, SW620 cells should be treated with TPA (100 ng/ml) to induce expression of MMP-7. Antibody should be titrated for optimal results in individual systems.

Legal Information

Manufactured by Daiichi Fine Chemical Co., Ltd. Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
IM37OP145AM80
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

species reactivity

human

species reactivity

human, rabbit, rat, guinea pig

species reactivity

human

species reactivity

human, Drosophila, rat, mouse

clone

141-7B2, monoclonal

clone

56-2A4, monoclonal

clone

4B2C1.11, monoclonal

clone

10H, monoclonal

Gene Information

human ... MMP7(4316)

Gene Information

human ... MMP9(4318)

Gene Information

human ... MDM2(4193)

Gene Information

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
IM40-100UG04055977220599

Questions

Reviews

No rating value

Active Filters