Skip to Content
Merck
All Photos(2)

Key Documents

HCYTMAG-60K-PX30

Millipore

MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel - Premixed 30 Plex - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Synonym(s):

Human cytokine multiplex kit, Luminex® human cytokine immunoassay, Millipore human cytokine panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 87-107%
standard curve range: 3.2-10,000 pg/mL

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

“Cytokine” is a general term used for a diverse group of soluble proteins and peptides which act as regulators under both normal and pathological conditions to modulate the functional activities of individual cells and tissues. These proteins also mediate direct interactions between cells and regulate processes taking place in the extracellular environment. Cytokines differ from hormones in that they act on a wider spectrum of target cells. Also, unlike hormones, they are not produced by specialized cells which are organized in specialized glands. The cytokine group of proteins includes lymphokines, interferons, colony stimulating factors and chemokines. Cytokine and chemokine research plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens, as well as disease states such as inflammatory disease, allergic reactions, irritable bowel disease (IBD), sepsis, and cancer.

The MILLIPLEX® Human Cytokine / Chemokine Panel 1 enables you to focus on the therapeutic potential of cytokines as well as the modulation of cytokine expression.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: EGF, G-CSF, GM-CSF, IFN-α2, IFN-γ, IL-1α, IL-1β, IL-1ra, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IP-10, MCP-1, MIP-1α, MIP-1β, TNF-α, TNF-β, VEGF, RANTES, Eotaxin/CCL11
  • Recommended Sample type: Serum, plasma, or tissue/cell lysate and culture supernatant samples
  • Recommended Sample dilution: Neat plasma or serum. Tissue culture supernatant may require a dilution with an appropriate control medium prior to assay. RANTES requires 1:100 dilution.
  • NOTE: RANTES cannot be combined with all other cytokines in this panel in serum/plasma due to a required 1:100 dilution
  • Assay Run Time: Overnight or two-hour primary incubation. For best results, an overnight incubation is recommended
  • Research Category: Inflammation & Immunology
  • Research Subcategory: Obesity, Metabolic Disorders, Inflammation & Autoimmune Mechanisms

Packaging

96 well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Note: RANTES, PDGF-AA and PDGF-AB/AA cannot be plexed with other analytes for serum/plasma.
Sensitivity: Refer to kit protocol for sensitivities for individual cytokines/chemokines.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergei Spitsin et al.
Journal of leukocyte biology, 101(4), 967-973 (2017-04-04)
The neuropeptide SP has physiologic and pathophysiologic roles in CNS and peripheral tissues and is involved in crosstalk between nervous and immune systems in various conditions, including HIV and SIV infection. Increased SP levels were demonstrated in plasma of HIV+
Farah Rahmatpanah et al.
Mucosal immunology, 12(1), 77-84 (2018-10-04)
Plasmacytoid dendritic cells (PDCs) are critical for defense against respiratory viruses because of their propensity to secrete high levels of type I interferons (IFN). The functions of PDCs in the lung can be influenced by airway epithelial cells. We examined
Wan-Ru Yu et al.
Biomedicines, 10(5) (2022-05-29)
Interstitial cystitis/bladder pain syndrome (IC/BPS) is a condition causing bladder inflammation. Urinary biomarkers have been assessed as suitable for the diagnosis and treatment. This study aimed at investigating the role of urinary biomarkers in identifying bladder conditions and predicting the
Catherine Laliberté et al.
Frontiers in oral health, 2, 740469-740469 (2022-01-21)
Background: Oral squamous cell carcinoma (OSCC) is a devastating disease that is usually associated with a dense associated inflammatory infiltrate. Characterizing tumor-associated inflammation is critical to understand the pathogenies of tumor development and progression. Methods: We have tested a protocol
Jeffrey S Barrett et al.
Journal of translational medicine, 14(1), 148-148 (2016-05-28)
Many HIV infected individuals with suppressed viral loads experience chronic immune activation frequently developing neurological impairment designated as HIV associated neurocognitive disorder (HAND). Adjunctive therapies may reduce HIV associated inflammation and therefore decrease the occurrence of HAND. We have conducted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service