Skip to Content
Merck
All Photos(1)

Key Documents

HCKP2-11K

Millipore

MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 - Immuno-Oncology Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

To order a Milliplex® kit, please search for your analyte of interest.

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Immune checkpoint proteins are regulators of immune activation and play a key role in maintaining immune homeostasis, preventing autoimmunity, and are involved in cancer immune escape. These immune system molecules demonstrate the ability to act both as agonists (co-stimulatory checkpoint molecules) or antagonists (co-inhibitory checkpoint molecules) of immune response. Immune checkpoint inhibitor drugs targeting inhibitory immune checkpoint proteins, such as CTLA-4 and PD-L1, have demonstrated efficacy in cancer immunotherapy by improving the antitumor immune response and saving lives in the treatment of many malignancies. Immune checkpoint proteins may express on immune cells and/or tumor cells as activating receptors or inhibitory receptors. Many immune checkpoint proteins are also expressed as soluble forms in circulation and in the tumor and tumor microenvironment and are putative biomarkers for immunotherapy in various diseases including cancer.MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 is a 31-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma and tissue culture samples: 4-1BBL/TNFSF9, 5′-NT/CD73 (5′-nucleotidase), APRIL (TNFSF13), Arginase-1, B7-H2/ICOSL (Inducible T cell costimulator ligand), B7-H3/CD276, B7-H4/VTCN1 (V-set domain-containing T-cell activation inhibitor 1), B7-H5/VISTA (V-set immunoregulatory receptor), B7-H6 (Natural killer cell cytotoxicity receptor 3 ligand), BAFF/BLyS (TNFSF13B), CD25/IL-2Rα (Interleukin-2 receptor alpha), CD30/TNFRSF8, CD40L (CD40 ligand), CD137/4-1BB (TNFRSF9), CD226/DNAM-1 (DNAX accessory molecule 1), E-Cadherin, FGL1/Hepassocin (Fibrinogen-like protein 1), Galectin-1 (Gal-1), Galectin-3 (Gal-3), Granulysin, Granzyme B, IDO1 (Indoleamine 2,3-dioxygenase 1), MICA (MHC class I polypeptide-related sequence A), MICB (MHC class I polypeptide-related sequence B), Nectin-2 (PVRL2, CD112), Nectin-4 (PVRL4), OX40/CD134 (TNFRSF4), Perforin, PVR/CD155 (Poliovirus receptor cell adhesion molecule), Siglec-7 (Sialic acid-binding Ig-like lectin 7, CD328), Siglec-9 (Sialic acid-binding Ig-like lectin 9, CD329).

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

The Human Immuno-Oncology Checkpoint Protein Panel 2 bead-based multiplex panel, using the Luminex® xMAP technology, enables the simultaneous analysis of 31 immune checkpoint proteins in human serum, plasma and cell culture samples.This is an overnight incubation assay (16-18 hours at 2-8°C).A maximum of 25 μL per well 1:2 diluted serum or plasma can be used. Tissue culture or other media may be run neat or appropriately diluted in control medium. Note: Best results using cell/tissue lysate samples may require diluting in assay buffer while testing similarly diluted lysis buffer for matrix effects.CD40 Ligand;4-1BBL/TNFSF9;Arginase-1;B7-H2/ICOSL;B7-H3/CD276;5′-NT/CD73;B7-H4/VTCN1;APRIL/TNFSF13;B7-H5/VISTA;CD25/IL-2Rα;B7-H6;CD137/4-1BB;Granzyme B;CD226/DNAM-1;CD30/TNFRSF8;E-Cadherin;FGL1/Hepassocin;Galectin-1 (GAL1);Galectin-3;Granulysin;IDO1;MICA;MICB;Nectin-2;BAFF/Blys;Nectin-4;OX40/CD134;PVR/CD155;Siglec-7;Siglec-9;Perforin

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Research evaluating immune checkpoints may help identify those likely to benefit from PD-1/PD-L1 immunotherapy and reveal new immunotherapy targets for future investigation. See how the MILLIPLEX® Human Immuno-Oncology Checkpoint Protein Panel 2 was used to multiplex immune checkpoint molecules for NSCLC research.

Cancer biomarker detection through MILLIPLEX® multiplex cancer assays can accelerate studies of tumorigenic processes helping advance cancer research.

Questions

1–2 of 2 Questions  
  1. What is the specificity of the Granulysin antibody pair in the Milliplex Human Immuno-oncology Checkpoint Protein Panel?

    1 answer
    1. The R&D team confirmed that both the capture and detection antibodies are polyclonal goat anti-human Granulysin. The immunogen is recombinant Human Granulysin (aa23-145). The antibodies have not been tested for reactivity to different isoforms of Granulysin.

      Helpful?

  2. How many samples can I analyze with one multiplex kit including all controls and assuming that I will have 5 different time points were I will need to analyze samples immediately?

    1 answer
    1. The end-user can analyze 38 samples in duplicates.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service