GR15
Anti-EGFR (Ab-5) Mouse mAb (EGFR.1)
liquid, clone EGFR.1, Calbiochem®
Synonym(s):
Anti-Epidermal Growth Factor Receptor
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
EGFR.1, monoclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
human
should not react with
rat, mouse
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG2b
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... EGFR(1956)
mouse ... Egfr(13649)
rat ... Egf(25313)
General description
Immunogen
Application
Flow Cytometry (1-5 g/ml)
Immunofluorescence (1-5 g/ml)
Immunoprecipitation (1-2 g/sample)
Neutralization Studies (1-2 g/ml; use Cat. No. GR15L; see application references)
Packaging
Warning
Physical form
Analysis Note
HL-60 cells
A-431 cells
Other Notes
Reynolds, F.H., Jr., et al. 1981. Nature292, 259.
Ushiro, H. and Cohen, S. 1980. J. Biol. Chem.255, 8363.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service