Skip to Content
Merck
All Photos(1)

Documents

GR13L

Sigma-Aldrich

Anti-EGFR (Ab-3) Mouse mAb (225)

lyophilized, clone 225, Calbiochem®

Synonym(s):

Anti-EGF-R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

225, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

should not react with

rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)
rat ... Egf(25313)

General description

Anti-EGFR (Ab-3), mouse monoclonal, clone 225 recognizes the ~170 kDa epidermal growth factor receptor protein. It is validated for use in immunoprecipitation and neutralization.
Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells (see application references). Recognizes the ~170 kDa epidermal growth factor receptor protein.
Recognizes the ~170 kDa epidermal growth factor receptor protein in A431 cells.

Immunogen

Human
partially purified, human EGFR from A431 cells

Application

Immunoprecipitation (1-2 µg per sample)

Neutralization Studies (2 µg/ml, see comments and application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 20 mM ammonium bicarbonate solution, 100 µg BSA.

Reconstitution

Reconstitute the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml. Lyophilized antibodies should be resuspended at 4°C with occasional gentle mixing for at least 2 h. Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate at 4°C with 0.1% azide for short-term storage. Avoid freeze/thaw cycles of solutions.

Analysis Note

Negative Control
HL-60 cells
Positive Control
A431 cells

Other Notes

Hunter T. 1984. Nature311, 414.
Reynolds, F.H., et. al 1981. Nature292, 259.
Ushiro, H. and Cohen, S. 1980. J. Biol. Chem.255, 8363.
This antibody has been shown to inhibit human tumor xenographs in nude mice. It inhibits EGF binding to its receptor and is an antagonist of in vivo EGF-stimulated tyrosine kinase activity. It also inhibits A431 tumor formation in nude mice. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pallavi Garg et al.
American journal of physiology. Lung cellular and molecular physiology, 301(1), L79-L90 (2011-05-03)
Thrombospondin-1 (TSP1) is a multidomain protein that contains epidermal growth factor (EGF)-like repeats that indirectly activate the EGF receptor (EGFR) and selected downstream signaling pathways. In these studies, we show that TSP1 opens the paracellular pathway in human lung microvascular
Anguo Liu et al.
The Journal of biological chemistry, 284(10), 6389-6402 (2009-01-09)
Thrombospondin (TSP) 1 is a trimeric multidomain protein that contains motifs that recognize distinct host cell receptors coupled to multiple signaling pathways. Selected TSP1-induced cellular responses are tyrosine kinase-dependent, and TSP1 contains epidermal growth factor (EGF)-like repeats. Specific receptor interactions
Caren C Bancroft et al.
International journal of cancer, 99(4), 538-548 (2002-05-07)
We previously reported that expression of angiogenesis factors interleukin-8 (IL-8) and vascular endothelial growth factor (VEGF) is promoted by coactivation of transcription factors nuclear factor-kappaB (NF-kappaB) and activator protein-1 (AP-1) by interleukin-1alpha in human head and neck squamous cell carcinomas
Silvia Ancona et al.
Biomedicines, 9(12) (2021-12-25)
Matrix metalloproteinase (MMP) dysregulation is implicated in several diseases, given their involvement in extracellular matrix degradation and cell motility. In lymphangioleiomyomatosis (LAM), a pulmonary rare disease, MMP-2 and MMP-9 have been detected at high levels in serum and urine. LAM
Jiarui Bi et al.
Journal of cell science, 133(5) (2019-11-15)
In periodontal disease (PD), bacterial biofilms cause gingival inflammation, leading to bone loss. In healthy individuals, αvβ6 integrin in junctional epithelium maintains anti-inflammatory transforming growth factor-β1 (TGF-β1) signaling, whereas its expression is lost in individuals with PD. Bacterial biofilms suppress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service