Skip to Content
Merck
All Photos(1)

Documents

CX2414

Supelco

Cyclopentane

OmniSolv®

Synonym(s):

Cyclopentane, Pentamethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
-CH2(CH2)3CH2-
UNSPSC Code:
41116105

Quality Level

product line

OmniSolv®

Assay

≥99.8%

form

liquid

composition

Cyclopentane, ≥85.0%
2,2-Dimethylbutane, ≤15.0%
n-pentane, ≤3.0%

impurities

≤0.02% Water (H2O)
≤5 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

transition temp

flash point 1.7 °C

UV cutoff

≤198 nm

UV absorption

λ: 198 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.50
λ: 220 nm Amax: ≤0.10
λ: 230 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

Application

OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (2,2-Dimethylbutane)15.0% maxAssay (Cyclopentane)85.0% minAssay (Total)99.8% minAssay (n-Pentane)3.0% maxCapillary ECD responsive substances (as PCNB)5ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)500ppt maxFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 198 nm1.00AU maxUV Abs. at 210 nm0.50AU maxUV Abs. at 220 nm0.10AU maxUV Abs. at 230 nm0.01AU maxUV Cut-off198nm maxWater (H2O)0.02% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.1 °F

Flash Point(C)

1.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service