Skip to Content
Merck
All Photos(1)

Key Documents

270601

Supelco

Cyclopentane

suitable for HPLC, ≥75% cyclopentane basis

Sign Into View Organizational & Contract Pricing

Select a Size

2 L
$946.00

$946.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 L
$946.00

About This Item

Empirical Formula (Hill Notation):
C5H10
CAS Number:
Molecular Weight:
70.13
Beilstein:
1900195
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
50 °C (lit.)
vapor pressure:
5.32 psi ( 20 °C)

$946.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

HPLC grade

Quality Level

vapor density

~2 (vs air)

vapor pressure

5.32 psi ( 20 °C)

Assay

≥75% cyclopentane basis

form

liquid

autoignition temp.

682 °F

expl. lim.

8.7 %

technique(s)

HPLC: suitable

evapn. residue

<0.0003%

refractive index

n20/D 1.405 (lit.)

bp

50 °C (lit.)

mp

−94 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 198 nm Amax: 1.0
λ: 210 nm Amax: 0.50
λ: 220 nm Amax: 0.10
λ: 240-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

C1CCCC1

InChI

1S/C5H10/c1-2-4-5-3-1/h1-5H2

InChI key

RGSFGYAAUTVSQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Glass distilled

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service