Skip to Content
Merck
All Photos(1)

Key Documents

BI0422

Sigma-Aldrich

Oxidizing Reagent, 0.02M I₂ in THF/Pyrid (89.6/0.4/10)

Biosynthesis Novabiochem®

Synonym(s):

DNA Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

liquid

composition

Iodine, 0.018-0.022 M
Pyridine, 0.36-0.44 % (v/v) GC
Tetrahydrofuran, 88.7-90.5 % (v/v) GC (solvent)
water, 9.0-11.0 % (v/v) GC

reaction suitability

reaction type: Oxidations
reagent type: oxidant

color

red

transition temp

flash point -13 °C

shipped in

ambient

storage temp.

room temp

Application

BI0422-0200 is packaged in 28/405 glass bottles
BI0422-0450 is packaged in 28/405 glass bottles
BI0422-0950 is packaged in 38/430 glass bottles
BI0422-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Assay (pyridine, GC): 0.36-0.44% v/v
Assay (tetrahydrofuran, GC): 88.7-90.5% v/v
Color: Red
Form: Liquid
Iodine Molarity: 0.018-0.022M
Water (H2O): 9.0-11.0% w/v

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F

Flash Point(C)

-13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service