Skip to Content
Merck
All Photos(1)

Key Documents

L060050

Sigma-Aldrich

Oxidizer

0.05 M, configured for (ÄKTA® and OligoPilot®)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$524.00
6 X 1 L
$1,290.00
4 X 2.5 L
$1,460.00

$524.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$524.00
6 X 1 L
$1,290.00
4 X 2.5 L
$1,460.00

About This Item

MDL number:
UNSPSC Code:
41116105
NACRES:
NA.21

$524.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

shelf life

180 days

concentration

11.4-14.0 g/L (Iodine content, titration)

color

clear

pH

8.5 (20 °C, 15 g/L)

bp

115 °C/1,013 hPa

mp

<-40 °C

compatibility

configured for (ÄKTA® and OligoPilot®)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Oxidizer solution, containing pyridine, iodine, and water, is applied to oxidize the chemically unstable trivalent phosphorous triester linkage formed in the coupling reaction to a stable pentavalent phosphate triester. Iodine functions as a mild oxidant and water functions as an oxygen donor.

Preparation Note

Pyridine / Water / Iodine 9/1/12,7 (v/v/w)

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service