Skip to Content
Merck
All Photos(1)

Key Documents

BDAM1

Millipore

mPAGE® Buffer Dam

for mPAGE® Mini Gel Tank

Synonym(s):

buffer adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105305
NACRES:
NC.01
Pricing and availability is not currently available.

packaging

pkg of 1 pack

manufacturer/tradename

Millipore

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

General description

mPAGE® Buffer Dam for use with mPAGE® Mini Gel Tank (MGT-2 and MGT-4) when running an odd number of gels.
The mPAGE® Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels. The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE® Gel Caster and mPAGE® Precast Gels. The mPAGE® Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.

Compatibility

Compatible with the mPAGE® Mini Gel Tank.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service